3-(4-Ethylphenyl)-5-isoxazolamine
Dodavatel: Apollo Scientific
Synonyms:
5-Amino-3-(4-ethylphenyl)isoxazole
OR470198-1G
OR470198-5G
APOSOR470198-1GEA
8700
CZK
APOSOR470198-1G
APOSOR470198-5G
3-(4-Ethylphenyl)-5-isoxazolamine
3-(4-Ethylphenyl)-5-isoxazolamine
Vzorec:
C₁₁H₁₂N₂O MW: 188,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07382879 CAS číslo: 1020955-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...