3-(3-Cyanophenoxy)propionic acid
Dodavatel: Apollo Scientific
OR470102-1G
OR470102-5G
APOSOR470102-1GEA
8700
CZK
APOSOR470102-1G
APOSOR470102-5G
3-(3-Cyanophenoxy)propionic acid
3-(3-Cyanophenoxy)propionic acid
Vzorec:
C₁₀H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09812167 CAS číslo: 210962-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...