2-Chloro-4-morpholinobenzenecarboxylic acid
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-4-morpholinobenzoic acid
OR470071-1G
OR470071-5G
APOSOR470071-1GEA
0
CZK
APOSOR470071-1G
APOSOR470071-5G
2-Chloro-4-morpholinobenzenecarboxylic acid
2-Chloro-4-morpholinobenzenecarboxylic acid
Vzorec:
C₁₁H₁₂ClNO₃ MW: 241,68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425732 CAS číslo: 175153-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...