Tisk…
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate
Dodavatel: Apollo Scientific
Synonyms: 3,3-Diethoxypropane-1,1-dicarboxylic Acid Diethyl Ester, Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate
OR470048-25G OR470048-5G
APOSOR470048-25GEA 26090 CZK
APOSOR470048-25G APOSOR470048-5G
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate
Vzorec: C₁₃H₂₄O₆
MW: 276.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671525
CAS číslo: 21339-47-9