Methyl-4-iodo-3-methylbenzoate 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-iodo-3-methylbenzoate
OR4700-5G
OR4700-100G
OR4700-25G
APOSOR4700-5GEA
827
CZK
APOSOR4700-5G
APOSOR4700-100G
APOSOR4700-25G
Methyl-4-iodo-3-methylbenzoate 97%
Methyl-4-iodo-3-methylbenzoate
Vzorec:
C₉H₉IO₂ MW: 276,07 g/mol Bod tání: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683863 CAS číslo: 5471-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...