Tisk…

Ethyl 3-Methylpyrazole-5-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 3-methyl-1H-pyrazole-5-carboxylate

OR4697-10G OR4697-1G OR4697-5G
APOSOR4697-10GEA 1450 CZK
APOSOR4697-10G APOSOR4697-1G APOSOR4697-5G
Ethyl 3-Methylpyrazole-5-carboxylate
Ethyl 3-Methylpyrazole-5-carboxylate
Vzorec: C₇H₁₀N₂O₂
MW: 154,165 g/mol
Bod tání: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00052514
CAS číslo: 4027-57-0
EINECS: 223-702-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR