Tisk…

Methyl-2-amino-5-iodobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-5-iodobenzoic acid methyll ester, Methyl 2-amino-5-iodobenzoate, Methyl 5-Iodoanthranilate, Methyl 2-Amino-5-iodobenzoate, 5-Iodoanthranilic acid methyll ester

OR4694-100G OR4694-25G OR4694-5G
APOSOR4694-100GEA 11680 CZK
APOSOR4694-100G APOSOR4694-25G APOSOR4694-5G
Methyl-2-amino-5-iodobenzoate
Methyl-2-amino-5-iodobenzoate
Vzorec: C₈H₈INO₂
MW: 277,0591 g/mol
Bod tání: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00798574
CAS číslo: 77317-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR