4-Bromobenzhydrol 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
p-Bromobenzhydrol, 4-Bromophenyl phenyl carbinol, 4-Bromodiphenylmethanol, 4-Bromo-α-phenylbenzyl alcohol, 4-Bromobenzhydryl alcohol, (4-Bromophenyl)(phenyl)methanol
OR4692-1G
OR4692-5G
OR4692-25G
APOSOR4692-1GEA
622
CZK
APOSOR4692-1G
APOSOR4692-5G
APOSOR4692-25G
4-Bromobenzhydrol 96%
4-Bromobenzhydrol
Vzorec:
C₁₃H₁₁BrO Bod tání: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016851 CAS číslo: 29334-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...