Methyl-4-amino-3-chlorobenzoate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 4-amino-3-chlorobenzoate
OR4674-1G
OR4674-5G
APOSOR4674-1GEA
622
CZK
APOSOR4674-1G
APOSOR4674-5G
Methyl-4-amino-3-chlorobenzoate 98%
Methyl-4-amino-3-chlorobenzoate
Vzorec:
C₈H₈ClNO₂ MW: 185,61 g/mol Bod tání: 107…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320686 CAS číslo: 84228-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...