5-Hydroxy-2,1,3-benzothiadiazole
Dodavatel: Apollo Scientific
Synonyms:
Benzo[c][1,2,5]thiadiazol-5-ol
OR46736-1G
OR46736-250MG
OR46736-500MG
APOSOR46736-1GEA
18250
CZK
APOSOR46736-1G
APOSOR46736-250MG
APOSOR46736-500MG
5-Hydroxy-2,1,3-benzothiadiazole
5-Hydroxy-2,1,3-benzothiadiazole
Vzorec:
C₆H₄N₂OS MW: 152,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00457955 CAS číslo: 768-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...