3-(2-Benzothiazolylthio)propionic acid
Dodavatel: Apollo Scientific
OR46731-1G
OR46731-5G
OR46731-10G
APOSOR46731-1GEA
662
CZK
APOSOR46731-1G
APOSOR46731-5G
APOSOR46731-10G
3-(2-Benzothiazolylthio)propionic acid
3-(2-Benzothiazolylthio)propionic acid
Vzorec:
C₁₀H₉NO₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00022878 CAS číslo: 4767-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...