3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid
Dodavatel: Apollo Scientific
Synonyms:
3,4-Dihydro-3-oxo-2H-(1,4)-benzothiazin-2-ylacetic acid, 2-(3-Oxo-3,4-dihydro-2H-benzo[b][1,4]thiazin-2-yl)acetic acid, 3-Oxo-3,4-dihydro-2H-1,4-benzothiazine-2-acetic acid
OR46730-10MG
OR46730-5MG
APOSOR46730-10MGEA
3110
CZK
APOSOR46730-10MG
APOSOR46730-5MG
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid
Vzorec:
C₁₀H₉NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD00087524 CAS číslo: 6270-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...