Tisk…

4-(1,3-Benzothiazol-2-yl)phenylamine

Dodavatel: Apollo Scientific

OR46725-250MG OR46725-500MG OR46725-2.5G OR46725-5G
APOSOR46725-250MGEA 2370 CZK
APOSOR46725-250MG APOSOR46725-500MG APOSOR46725-2.5G APOSOR46725-5G
4-(1,3-Benzothiazol-2-yl)phenylamine
4-(1,3-Benzothiazol-2-yl)phenylamine
Vzorec: C₁₃H₁₀N₂S
Storage Temperature: Ambient
MDL Number: MFCD00182896
CAS číslo: 6278-73-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR