1-Isobutyl-1H-pyrrole
Dodavatel: Apollo Scientific
OR46713-1G
OR46713-250MG
OR46713-500MG
APOSOR46713-1GEA
9690
CZK
APOSOR46713-1G
APOSOR46713-250MG
APOSOR46713-500MG
1-Isobutyl-1H-pyrrole
1-Isobutyl-1H-pyrrole
Vzorec:
C₈H₁₃N Storage Temperature: Ambient |
MDL Number:
MFCD12187092 CAS číslo: 20884-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...