1-Pyridin-2-ylcyclopropanecarbonitrile
Dodavatel: Apollo Scientific
OR46709-1G
OR46709-250MG
OR46709-500MG
OR46709-2.5G
APOSOR46709-1GEA
11010
CZK
APOSOR46709-1G
APOSOR46709-250MG
APOSOR46709-500MG
APOSOR46709-2.5G
1-Pyridin-2-ylcyclopropanecarbonitrile
1-Pyridin-2-ylcyclopropanecarbonitrile
Vzorec:
C₉H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD15526928 CAS číslo: 162960-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...