1-(4-Methylpyridin-3-yl)ethanone
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(4-Methyl-3-pyridinyl)-Ethanone
OR46708-1G
OR46708-250MG
OR46708-500MG
APOSOR46708-1GEA
2610
CZK
APOSOR46708-1G
APOSOR46708-250MG
APOSOR46708-500MG
1-(4-Methylpyridin-3-yl)ethanone
1-(4-Methylpyridin-3-yl)ethanone
Vzorec:
C₈H₉NO MW: 135.17 g/mol Bod varu: 57…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD08447082 CAS číslo: 51227-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...