2-Hydroxybenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR46678-1G
OR46678-250MG
OR46678-500MG
APOSOR46678-1GEA
17490
CZK
APOSOR46678-1G
APOSOR46678-250MG
APOSOR46678-500MG
2-Hydroxybenzoyl chloride
2-Hydroxybenzoyl chloride
Vzorec:
C₇H₅ClO₂ Bod tání: 17…20 °C Storage Temperature: Ambient |
MDL Number:
MFCD11169645 CAS číslo: 1441-87-8 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...