2-Mercaptobenzoyl chloride
Dodavatel: Apollo Scientific
OR46677-1G
OR46677-250MG
OR46677-500MG
APOSOR46677-1GEA
14520
CZK
APOSOR46677-1G
APOSOR46677-250MG
APOSOR46677-500MG
2-Mercaptobenzoyl chloride
2-Mercaptobenzoyl chloride
Vzorec:
C₇H₅ClOS Storage Temperature: Ambient |
MDL Number:
MFCD22054937 CAS číslo: 1677-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...