Tisk…

2-(Phenylamino)acetyl chloride

Dodavatel: Apollo Scientific

OR46674-1G OR46674-250MG OR46674-500MG OR46674-2.5G
APOSOR46674-1GEA 14520 CZK
APOSOR46674-1G APOSOR46674-250MG APOSOR46674-500MG APOSOR46674-2.5G
2-(Phenylamino)acetyl chloride
2-(Phenylamino)acetyl chloride
Vzorec: C₈H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD20661775
CAS číslo: 85630-85-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR