Tisk…

4-(Morpholin-4-yl)benzoyl chloride hydrochloride

Dodavatel: Apollo Scientific

OR46670-1G OR46670-250MG OR46670-500MG
APOSOR46670-1GEA 14780 CZK
APOSOR46670-1G APOSOR46670-250MG APOSOR46670-500MG
4-(Morpholin-4-yl)benzoyl chloride hydrochloride
4-(Morpholin-4-yl)benzoyl chloride hydrochloride
Vzorec: C₁₁H₁₃Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD08448240
CAS číslo: 162848-18-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR