Tisk…

2-Oxocyclohexanecarboxylic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR46664-1G OR46664-250MG
APOSOR46664-1GEA 8900 CZK
APOSOR46664-1G APOSOR46664-250MG
2-Oxocyclohexanecarboxylic acid 97%
2-Oxocyclohexanecarboxylic acid
Vzorec: C₇H₁₀O₃
Bod tání: 79…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797988
CAS číslo: 18709-01-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR