Tisk…

1,2,3,4-Tetrahydroquinoline-4-carboxylic acid

Dodavatel: Apollo Scientific

OR46659-1G OR46659-50MG OR46659-100MG OR46659-250MG OR46659-500MG OR46659-2.5G
APOSOR46659-1GEA 9330 CZK
APOSOR46659-1G APOSOR46659-50MG APOSOR46659-100MG APOSOR46659-250MG APOSOR46659-500MG APOSOR46659-2.5G
1,2,3,4-Tetrahydroquinoline-4-carboxylic acid
1,2,3,4-Tetrahydroquinoline-4-carboxylic acid
Vzorec: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834787
CAS číslo: 13337-69-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR