Tisk…

3-(Pyrrolidin-1-ylmethyl)benzoic acid

Dodavatel: Apollo Scientific

OR46652-1G OR46652-250MG OR46652-500MG OR46652-5G
APOSOR46652-1GEA 13030 CZK
APOSOR46652-1G APOSOR46652-250MG APOSOR46652-500MG APOSOR46652-5G
3-(Pyrrolidin-1-ylmethyl)benzoic acid
3-(Pyrrolidin-1-ylmethyl)benzoic acid
Vzorec: C₁₂H₁₅NO₂
MW: 205.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05841592
CAS číslo: 884507-41-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR