3-Hydroxytetrahydropyran
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Hydroxy-2H-pyran, Tetrahydro-2H-pyran-3-ol
OR46583-1G
OR46583-250MG
APOSOR46583-1GEA
744
CZK
APOSOR46583-1G
APOSOR46583-250MG
3-Hydroxytetrahydropyran
3-Hydroxytetrahydropyran
Vzorec:
C₅H₁₀O₂ MW: 102,1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00219387 CAS číslo: 19752-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...