Tisk…

3-(1,2-Dihydro-4-oxopyrrolo-[3,2,1-ij]quinolin-5-yl)-(2E)-propenoic acid 95+%

Dodavatel: Apollo Scientific

OR4658-10G OR4658-1G OR4658-5G
APOSOR4658-10GEA 27250 CZK
APOSOR4658-10G APOSOR4658-1G APOSOR4658-5G
3-(1,2-Dihydro-4-oxopyrrolo-[3,2,1-ij]quinolin-5-yl)-(2E)-propenoic acid 95+%
3-(1,2-Dihydro-4-oxopyrrolo-[3,2,1-ij]quinolin-5-yl)-(2E)-propenoic acid
Vzorec: C₁₄H₁₁NO₃
Bod tání: >265 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD01567774
CAS číslo: 386715-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR