1,4-Dioxaspiro[4.5]decan-8-ol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Hydroxycyclohexan-1-one monoethylene ketal, 4-Hydroxycyclohexanone ethylene acetal
OR46567-25G
OR46567-100G
OR46567-5G
APOSOR46567-25GEA
2280
CZK
APOSOR46567-25G
APOSOR46567-100G
APOSOR46567-5G
1,4-Dioxaspiro[4.5]decan-8-ol 98%
1,4-Dioxaspiro[4.5]decan-8-ol
Vzorec:
C₈H₁₄O₃ MW: 158,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067003 CAS číslo: 22428-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...