5-(4-Bromophenyl)-4,6-dichloropyrimidine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR46538-25G
OR46538-5G
OR46538-1G
APOSOR46538-25GEA
26920
CZK
APOSOR46538-25G
APOSOR46538-5G
APOSOR46538-1G
5-(4-Bromophenyl)-4,6-dichloropyrimidine 98%
5-(4-Bromophenyl)-4,6-dichloropyrimidine
Vzorec:
C₁₀H₅BrCl₂N₂ MW: 303,97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13151897 CAS číslo: 146533-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...