Chelidonic acid monohydrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Oxo-4H-pyran-2,6-dicarboxylic acid monohydrate, 2,6-Dicarboxy-4-oxo-4H-pyran monohydrate
OR46508-100G
OR46508-25G
OR46508-5G
APOSOR46508-100GEA
20010
CZK
APOSOR46508-100G
APOSOR46508-25G
APOSOR46508-5G
Chelidonic acid monohydrate
Chelidonic acid monohydrate
Vzorec:
C₇H₄O₆.H₂O Bod tání: 265 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149433 CAS číslo: 6003-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...