Tisk…

2,5-Diethoxythiophenol

Dodavatel: Apollo Scientific
Varování

OR4650-100G OR4650-25G OR4650-5G
APOSOR4650-100GEA 21230 CZK
APOSOR4650-100G APOSOR4650-25G APOSOR4650-5G
2,5-Diethoxythiophenol
2,5-Diethoxythiophenol
Vzorec: C₁₀H₁₄O₂S
Storage Temperature: Ambient
CAS číslo: 29236-93-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR