Tisk…

5-Bromo-2-iodoanisole 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Bromo-1-iodo-2-methoxybenzene

OR46223-1G OR46223-25G OR46223-5G
APOSOR46223-1GEA 992 CZK
APOSOR46223-1G APOSOR46223-25G APOSOR46223-5G
5-Bromo-2-iodoanisole 97%
5-Bromo-2-iodoanisole
Vzorec: C₇H₆BrIO
MW: 312.9300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847482
CAS číslo: 791642-68-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR