Tisk…

2-Aminopyridin-4-ol 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Amino-4-hydroxypyridine

OR46212-25G OR46212-100G
APOSOR46212-25GEA 2240 CZK
APOSOR46212-25G APOSOR46212-100G
2-Aminopyridin-4-ol 95%
2-Aminopyridin-4-ol
Vzorec: C₅H₆N₂O
MW: 110,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114161
CAS číslo: 33631-05-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR