(S)-2-Methylpyrrolidine 95%
Dodavatel: Apollo Scientific
OR4618-25G
OR4618-5G
APOSOR4618-25GEA
26320
CZK
APOSOR4618-25G
APOSOR4618-5G
(S)-2-Methylpyrrolidine 95%
(S)-2-Methylpyrrolidine
Vzorec:
C₅H₁₁N MW: 85,1475 g/mol Bod varu: 97…99 °C Hustota: 0,834 g/cm³ (25 °C) Bod vzplanutí: 7,2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07779425 CAS číslo: 59335-84-1 UN: 2924 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...