Tisk…

2,6-Diiodopyrazine

Dodavatel: Apollo Scientific

OR46149-1G OR46149-5G
APOSOR46149-1GEA 0 CZK
APOSOR46149-1G APOSOR46149-5G
2,6-Diiodopyrazine
2,6-Diiodopyrazine
Vzorec: C₄H₂I₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD09265492
CAS číslo: 58138-79-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR