(-)-Corey lactone-4-phenylbenzoate 98%
Dodavatel: Apollo Scientific
Synonyms:
(3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl 4-Phenylbenzoate, (3aR,4S,5R,6aS)-Hexahydro-4-(hydroxymethyl)-5-(4-phenylbenzoyloxy)-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Biphenyl-4-carboxylate
OR46130-5G
APOSOR46130-5GEA
3810
CZK
APOSOR46130-5G
(-)-Corey lactone-4-phenylbenzoate 98%
(-)-Corey lactone-4-phenylbenzoate
Vzorec:
C₂₁H₂₀O₅ MW: 352,39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078077 CAS číslo: 31752-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...