Tisk…

1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid 95+%

Dodavatel: Apollo Scientific

Synonyms: 7-Azaindole-2-carboxylic acid

OR46104-1G OR46104-5G
APOSOR46104-1GEA 1700 CZK
APOSOR46104-1G APOSOR46104-5G
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid 95+%
1H-Pyrrolo[2,3-b]pyridine-2-carboxylic acid
Vzorec: C₈H₆N₂O₂
MW: 162,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659752
CAS číslo: 136818-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR