1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Azaindole-3-carboxaldehyde
OR46103-1G
OR46103-5G
OR46103-10G
APOSOR46103-1GEA
2530
CZK
APOSOR46103-1G
APOSOR46103-5G
APOSOR46103-10G
1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde 97%
1H-Pyrrolo[3,2-b]pyridine-3-carbaldehyde
Vzorec:
C₈H₆N₂O MW: 146,1460 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11877845 CAS číslo: 276862-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...