Tisk…

Tridecanol (mixture of isomers)

Dodavatel: Apollo Scientific
Varování

Synonyms: Tridecyl Alcohol

OR46094-10G OR46094-1G
APOSOR46094-10GEA 705 CZK
APOSOR46094-10G APOSOR46094-1G
Tridecanol (mixture of isomers)
Tridecanol (mixture of isomers)
Vzorec: C₁₃H₂₈O
Bod varu: 155…156 °C/15mmHg
Bod tání: 29…34 °C
Hustota: 0,822 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01778419
CAS číslo: 26248-42-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR