3-Methylcrotonic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid
OR46069-25G
OR46069-100G
APOSOR46069-25GEA
622
CZK
APOSOR46069-25G
APOSOR46069-100G
3-Methylcrotonic acid 98%
3-Methylcrotonic acid
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 194…195 °C Bod tání: 65…70 °C Hustota: 1,969 g/cm³ (25 °C) Bod vzplanutí: 93,4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004366 CAS číslo: 541-47-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...