Tisk…

6-Hydroxy-2,4,5-triaminopyrimidine 95%

Dodavatel: Apollo Scientific
Varování

OR46053-5G OR46053-25G
APOSOR46053-5GEA 3740 CZK
APOSOR46053-5G APOSOR46053-25G
6-Hydroxy-2,4,5-triaminopyrimidine 95%
6-Hydroxy-2,4,5-triaminopyrimidine
Vzorec: C₄H₇N₅O
Storage Temperature: Ambient
MDL Number: MFCD00217058
CAS číslo: 1004-75-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR