Tisk…

Diethyl dibromomalonate

Dodavatel: Apollo Scientific
Nebezpečí

OR46029-100G OR46029-25G
APOSOR46029-100GEA 2570 CZK
APOSOR46029-100G APOSOR46029-25G
Diethyl dibromomalonate
Diethyl dibromomalonate
Vzorec: C₇H₁₀Br₂O₄
Bod varu: 108…110 °C/4mmHg
Hustota: 1,68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015154
CAS číslo: 631-22-1
EINECS: 211-151-9
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR