2-Nitrobenzyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(Chloromethyl)-2-nitrobenzene, 2-Nitrobenzyl chloride, α-Chloro-o-nitrotoluene, alpha-Chloro-2-nitrotoluene
OR46019-5G
OR46019-1G
APOSOR46019-5GEA
2030
CZK
APOSOR46019-5G
APOSOR46019-1G
2-Nitrobenzyl chloride 98%
2-Nitrobenzyl chloride
Vzorec:
C₇H₆ClNO₂ MW: 171,58 g/mol Bod varu: 127…133 °C/10mmHg Bod tání: 46…48 °C Bod vzplanutí: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007185 CAS číslo: 612-23-7 EINECS: 210-300-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...