2-Iodobenzoyl chloride
Dodavatel: Apollo Scientific
OR460054-25G
OR460054-5G
APOSOR460054-25GEA
1880
CZK
APOSOR460054-25G
APOSOR460054-5G
2-Iodobenzoyl chloride
2-Iodobenzoyl chloride
Vzorec:
C₇H₄ClIO Storage Temperature: Ambient |
MDL Number:
MFCD00001040 CAS číslo: 609-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...