3-Iodobenzoyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR460051-25G
OR460051-5G
APOSOR460051-25GEA
9760
CZK
APOSOR460051-25G
APOSOR460051-5G
3-Iodobenzoyl chloride
3-Iodobenzoyl chloride
Vzorec:
C₇H₄ClIO Bod varu: 159…160 °C/23mmHg Bod tání: 23…25 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093934 CAS číslo: 1711-10-0 EINECS: 216-979-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...