Tisk…

2-Aminopyrimidine-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR46000-1G OR46000-250MG
APOSOR46000-1GEA 2690 CZK
APOSOR46000-1G APOSOR46000-250MG
2-Aminopyrimidine-4-carboxylic acid
2-Aminopyrimidine-4-carboxylic acid
Vzorec: C₅H₅N₃O₂
MW: 139,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783936
CAS číslo: 2164-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR