1-(6-Methylpyridin-2-yl)-2-methylsulphonylethanone 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methyl-6-[(methylsulphonyl)acetyl]pyridine
OR4562-10G
OR4562-5G
APOSOR4562-10GEA
12400
CZK
APOSOR4562-10G
APOSOR4562-5G
1-(6-Methylpyridin-2-yl)-2-methylsulphonylethanone 95+%
1-(6-Methylpyridin-2-yl)-2-methylsulphonylethanone
Vzorec:
C₉H₁₁NO₃S MW: 213.26 g/mol Bod tání: 65…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD01941311 CAS číslo: 386715-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...