3-Bromobenzyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Bromo-α-chlorotoluene, 1-(Chloromethyl)-3-bromobenzene, α-Chloro-3-bromotoluene, 1-Bromo-3-(chloromethyl)benzene
OR4547-25G
OR4547-100G
APOSOR4547-25GEA
2200
CZK
APOSOR4547-25G
APOSOR4547-100G
3-Bromobenzyl chloride
3-Bromobenzyl chloride
Vzorec:
C₇H₆BrCl MW: 205,48 g/mol Bod varu: 118…119 °C/18mmHg Hustota: 1,608 g/cm³ (25 °C) Bod vzplanutí: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040865 CAS číslo: 932-77-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...