Tisk…

3-Chloro-N-methylbenzylamine 97%

Dodavatel: Apollo Scientific

Synonyms: 1-(3-Chlorophenyl)-N-methylmethanamine, N-Methyl-3-chlorobenzylamine

OR452216-25G OR452216-5G
APOSOR452216-25GEA 7580 CZK
APOSOR452216-25G APOSOR452216-5G
3-Chloro-N-methylbenzylamine 97%
3-Chloro-N-methylbenzylamine
Vzorec: C₈H₁₀ClN
MW: 155,625 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045194
CAS číslo: 39191-07-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR