Tisk…

1-(Dimethylamino)isopropylamine 97%

Dodavatel: Apollo Scientific

Synonyms: N1,N1-Dimethyl-1,2-propanediamine dihydrochloride

OR452189-1G OR452189-500MG
APOSOR452189-1GEA 5590 CZK
APOSOR452189-1G APOSOR452189-500MG
1-(Dimethylamino)isopropylamine 97%
1-(Dimethylamino)isopropylamine
Vzorec: C₅H₁₄N₂
MW: 102,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014812
CAS číslo: 108-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR