Tisk…

1-Benzoyl-2-piperidinecarboxylic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Benzoylpiperidine-2-carboxylic acid

OR452128-1G OR452128-5G
APOSOR452128-1GEA 3440 CZK
APOSOR452128-1G APOSOR452128-5G
1-Benzoyl-2-piperidinecarboxylic acid 97%
1-Benzoyl-2-piperidinecarboxylic acid
Vzorec: C₁₃H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD02211013
CAS číslo: 78348-46-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR