Tisk…

(1-Methylpiperidin-4-yl)acetic acid hydrochloride 97%

Dodavatel: Apollo Scientific

OR452122-1G OR452122-500MG
APOSOR452122-1GEA 14880 CZK
APOSOR452122-1G APOSOR452122-500MG
(1-Methylpiperidin-4-yl)acetic acid hydrochloride 97%
(1-Methylpiperidin-4-yl)acetic acid hydrochloride
Vzorec: C₈H₁₆ClNO₂
MW: 193,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12196931
CAS číslo: 67686-06-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR